Assay Details
Gene Symbol: NDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345216.1 | NP_001330516.1 | 9 - 10 | 1062 | 73 | |
NM_125058.3 | NP_200486.1 | 9 - 10 | 1178 | 73 | ||
GenBank mRNA | AY050428.1 | - | 9 - 10 | 930 | 73 | |
AY087544.1 | - | 9 - 10 | 930 | 73 | ||
AY090332.1 | - | 9 - 10 | 821 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
