Assay Details
Gene Symbol: AT5G56840 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125067.4 | NP_200495.1 | 2 - 3 | 626 | 97 | |
GenBank mRNA | AY519517.1 | - | 2 - 3 | 475 | 97 | |
BT012528.1 | - | 2 - 3 | 571 | 97 | ||
BT014956.1 | - | 2 - 3 | 475 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
