Assay Details
Gene Symbol: MYB36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125143.4 | NP_200570.1 | 1 - 2 | 275 | 71 | |
GenBank mRNA | AK228818.1 | - | 1 - 2 | 217 | 71 | |
AY519640.1 | - | 1 - 2 | 138 | 71 | ||
BT030331.1 | - | 1 - 2 | 138 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
