Assay Details
Gene Symbol: MYB36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125143.4 | NP_200570.1 | 2 - 3 | 405 | 140 | |
GenBank mRNA | AK228818.1 | - | 2 - 3 | 347 | 140 | |
AY519640.1 | - | 2 - 3 | 268 | 140 | ||
BT030331.1 | - | 2 - 3 | 268 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
