Assay Details
Gene Symbol: AT5G58400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125226.4 | NP_200648.1 | 1 - 2 | 308 | 82 | |
GenBank mRNA | AK118274.1 | - | 1 - 2 | 281 | 82 | |
BT008527.1 | - | 1 - 2 | 230 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
