Assay Details
Gene Symbol: AT5G58400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125226.4 | NP_200648.1 | 2 - 3 | 492 | 103 | |
GenBank mRNA | AK118274.1 | - | 2 - 3 | 465 | 103 | |
BT008527.1 | - | 2 - 3 | 414 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
