Assay Details
Gene Symbol: PLC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345317.1 | NP_001332555.1 | 4 - 5 | 1112 | 117 | |
NM_001345318.1 | NP_001332556.1 | 4 - 5 | 805 | 117 | ||
NM_125256.3 | NP_200677.2 | 3 - 4 | 1186 | 117 | ||
GenBank mRNA | AF434167.2 | - | 3 - 4 | 749 | 117 | |
AY062681.1 | - | 3 - 4 | 759 | 117 | ||
BT010399.1 | - | 3 - 4 | 658 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
