Assay Details
Gene Symbol: PRR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203645.2 | NP_001190574.1 | 5 - 6 | 1180 | 81 | |
NM_001203646.1 | NP_001190575.1 | 4 - 5 | 1217 | 81 | ||
NM_001345386.1 | NP_001331790.1 | 4 - 5 | 1653 | 81 | ||
NM_001345387.1 | NP_001331791.1 | 4 - 5 | 1653 | 81 | ||
NM_001345388.1 | NP_001331792.1 | 5 - 6 | 1176 | 81 | ||
NM_001345389.1 | NP_001331789.1 | 5 - 6 | 1129 | 81 | ||
NM_125403.3 | NP_568919.1 | 5 - 6 | 1177 | 81 | ||
GenBank mRNA | AB046956.1 | - | 4 - 5 | 740 | 81 | |
AB493801.1 | - | 4 - 5 | 740 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
