Assay Details
Gene Symbol: AT5G61620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125556.2 | NP_200970.1 | 1 - 2 | 382 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
