Assay Details
Gene Symbol: MES5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121068.4 | NP_196592.1 | 2 - 3 | 720 | 132 | |
GenBank mRNA | AK226255.1 | - | 2 - 3 | 556 | 132 | |
AY093714.1 | - | 2 - 3 | 491 | 132 | ||
AY142490.1 | - | 2 - 3 | 491 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
