Assay Details
Gene Symbol: NDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343214.1 | NP_001332289.1 | 10 - 11 | 1791 | 73 | |
NM_121217.3 | NP_568251.1 | 10 - 11 | 1093 | 73 | ||
GenBank mRNA | AF361823.1 | - | 10 - 11 | 947 | 73 | |
AY059161.1 | - | 10 - 11 | 819 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
