Assay Details
Gene Symbol: NDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121217.3 | NP_568251.1 | 1 - 2 | 333 | 83 | |
GenBank mRNA | AF361823.1 | - | 1 - 2 | 187 | 83 | |
AY059161.1 | - | 1 - 2 | 59 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
