Assay Details
Gene Symbol: NDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343214.1 | NP_001332289.1 | 2 - 3 | 1135 | 147 | |
NM_121217.3 | NP_568251.1 | 2 - 3 | 437 | 147 | ||
GenBank mRNA | AF361823.1 | - | 2 - 3 | 291 | 147 | |
AY059161.1 | - | 2 - 3 | 163 | 147 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001343214.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
