Assay Details
Gene Symbol: CKS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128355.3 | NP_180363.1 | 2 - 3 | 375 | 93 | |
GenBank mRNA | AJ000016.1 | - | 2 - 3 | 131 | 93 | |
AK175719.1 | - | 2 - 3 | 186 | 93 | ||
AK175828.1 | - | 2 - 3 | 200 | 93 | ||
AK176119.1 | - | 2 - 3 | 186 | 93 | ||
AY088339.1 | - | 2 - 3 | 201 | 93 | ||
BT024528.1 | - | 2 - 3 | 116 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
