Assay Details
Gene Symbol: RAP2.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202696.2 | NP_001189625.1 | 4 - 5 | 1035 | 139 | |
NM_001336168.1 | NP_001318304.1 | 4 - 5 | 1035 | 139 | ||
NM_001336169.1 | NP_001323947.1 | 4 - 5 | 1035 | 139 | ||
NM_001336170.1 | NP_001323946.1 | 3 - 4 | 1068 | 139 | ||
NM_001336171.1 | NP_001323945.1 | 4 - 5 | 954 | 139 | ||
NM_128415.5 | NP_565674.1 | 4 - 5 | 1035 | 139 | ||
GenBank mRNA | AF325074.1 | - | 4 - 5 | 662 | 139 | |
AY050815.1 | - | 4 - 5 | 952 | 139 | ||
AY091418.1 | - | 4 - 5 | 662 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
