Assay Details
Gene Symbol: RAP2.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336170.1 | NP_001323946.1 | 4 - 5 | 1204 | 72 | |
NM_128415.5 | NP_565674.1 | 5 - 6 | 1171 | 72 | ||
GenBank mRNA | AF325074.1 | - | 5 - 6 | 798 | 72 | |
AY050815.1 | - | 5 - 6 | 1088 | 72 | ||
AY091418.1 | - | 5 - 6 | 798 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
