Assay Details
Gene Symbol: RAP2.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202696.2 | NP_001189625.1 | 7 - 8 | 1435 | 86 | |
NM_001336168.1 | NP_001318304.1 | 5 - 6 | 1648 | 86 | ||
NM_001336169.1 | NP_001323947.1 | 6 - 7 | 1542 | 86 | ||
NM_001336170.1 | NP_001323946.1 | 5 - 6 | 1423 | 86 | ||
NM_001336171.1 | NP_001323945.1 | 6 - 7 | 1460 | 86 | ||
NM_128415.5 | NP_565674.1 | 6 - 7 | 1390 | 86 | ||
GenBank mRNA | AF325074.1 | - | 6 - 7 | 1017 | 86 | |
AY050815.1 | - | 6 - 7 | 1307 | 86 | ||
AY091418.1 | - | 6 - 7 | 1017 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
