Assay Details
Gene Symbol: AT2G28660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128427.3 | NP_180434.2 | 1 - 2 | 711 | 76 | |
GenBank mRNA | AY303948.1 | - | 1 - 2 | 547 | 76 | |
BT029348.1 | - | 1 - 2 | 547 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
