Assay Details
Gene Symbol: AT2G28660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128427.3 | NP_180434.2 | 2 - 3 | 783 | 86 | |
GenBank mRNA | AY303948.1 | - | 2 - 3 | 619 | 86 | |
BT029348.1 | - | 2 - 3 | 619 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
