Assay Details
Gene Symbol: TWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113059.3 | NP_188801.2 | 5 - 6 | 644 | 95 | |
GenBank mRNA | AJ224640.1 | - | 5 - 6 | 644 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
