Assay Details
Gene Symbol: AT3G22250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113123.2 | NP_188864.1 | 1 - 2 | 523 | 74 | |
GenBank mRNA | KJ138813.1 | - | 1 - 2 | 470 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
