Assay Details
Gene Symbol: AT3G22250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113123.2 | NP_188864.1 | 2 - 3 | 1180 | 75 | |
GenBank mRNA | KJ138813.1 | - | 2 - 3 | 1127 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
