Assay Details
Gene Symbol: AT3G23310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113233.5 | NP_188973.2 | 11 - 12 | 1936 | 122 | |
GenBank mRNA | AY063969.1 | - | 11 - 12 | 1770 | 122 | |
AY096444.1 | - | 10 - 11 | 1404 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
