Assay Details
Gene Symbol: AAE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113283.4 | NP_189021.2 | 6 - 7 | 1092 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AAE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113283.4 | NP_189021.2 | 6 - 7 | 1092 | 87 |
Set Membership: |
![]() ![]() ![]() ![]() |