Assay Details
Gene Symbol: MSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113339.4 | NP_189075.2 | 4 - 5 | 585 | 108 | |
GenBank mRNA | AY191303.1 | - | 4 - 5 | 551 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
