Assay Details
Gene Symbol: RPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113496.5 | NP_189221.1 | 2 - 3 | 543 | 115 | |
GenBank mRNA | AF370235.1 | - | 2 - 3 | 471 | 115 | |
AF424601.1 | - | 2 - 3 | 471 | 115 | ||
AY059125.1 | - | 2 - 3 | 446 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
