Assay Details
Gene Symbol: RGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338780.1 | NP_001327634.1 | 6 - 7 | 971 | 67 | |
NM_113514.4 | NP_189238.2 | 6 - 7 | 971 | 67 | ||
GenBank mRNA | AY150429.1 | - | 6 - 7 | 701 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
