Assay Details
Gene Symbol: AT5G64100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125806.3 | NP_201215.1 | 1 - 2 | 611 | 80 | |
GenBank mRNA | X98808.1 | - | 481 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
