Assay Details
Gene Symbol: AT5G64110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125807.5 | NP_201216.1 | 1 - 2 | 540 | 67 | |
GenBank mRNA | AY065173.1 | - | 1 - 2 | 501 | 67 | |
BT003353.1 | - | 1 - 2 | 419 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
