Assay Details
Gene Symbol: AT5G64120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125808.3 | NP_201217.1 | 1 - 2 | 479 | 87 | |
GenBank mRNA | AF428274.1 | - | 1 - 2 | 475 | 87 | |
BT002622.1 | - | 1 - 2 | 418 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
