Assay Details
Gene Symbol: AT5G64120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125808.3 | NP_201217.1 | 2 - 3 | 643 | 77 | |
GenBank mRNA | AF428274.1 | - | 2 - 3 | 639 | 77 | |
BT002622.1 | - | 2 - 3 | 582 | 77 | ||
X99097.1 | - | 734 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
