Assay Details
Gene Symbol: XRCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345635.1 | NP_001318876.1 | 8 - 9 | 1067 | 97 | |
NM_125848.3 | NP_201257.2 | 8 - 9 | 1078 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
