Assay Details
Gene Symbol: DPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125884.4 | NP_201291.1 | 15 - 16 | 1723 | 80 | |
GenBank mRNA | AY037231.1 | - | 15 - 16 | 1652 | 80 | |
AY081744.1 | - | 15 - 16 | 1632 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
