Assay Details
Gene Symbol: DPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125884.4 | NP_201291.1 | 3 - 4 | 597 | 83 | |
GenBank mRNA | AY037231.1 | - | 3 - 4 | 526 | 83 | |
AY081744.1 | - | 3 - 4 | 506 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
