Assay Details
Gene Symbol: AT1G11320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101005.4 | NP_172599.1 | 1 - 2 | 527 | 119 | |
GenBank mRNA | AK228636.1 | - | 1 - 2 | 213 | 119 | |
BT005740.1 | - | 1 - 2 | 213 | 119 | ||
BT006093.1 | - | 1 - 2 | 108 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
