Assay Details
Gene Symbol: AT1G11320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101005.4 | NP_172599.1 | 2 - 3 | 994 | 76 | |
GenBank mRNA | AK228636.1 | - | 2 - 3 | 680 | 76 | |
BT005740.1 | - | 2 - 3 | 680 | 76 | ||
BT006093.1 | - | 2 - 3 | 575 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
