Assay Details
Gene Symbol: AT1G12300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101102.3 | NP_172694.1 | 1 | 831 | 75 | |
GenBank mRNA | AK230459.1 | - | 1 | 709 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
