Assay Details
Gene Symbol: GAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084061.1 | NP_001077530.1 | 10 - 11 | 954 | 80 | |
NM_101214.4 | NP_172801.1 | 10 - 11 | 1269 | 80 | ||
GenBank mRNA | AF410271.1 | - | 10 - 11 | 956 | 80 | |
AY049259.1 | - | 10 - 11 | 956 | 80 | ||
AY090275.1 | - | 9 - 10 | 885 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
