Assay Details
Gene Symbol: DWA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106276.5 | NP_177753.1 | 10 - 11 | 1047 | 122 | |
GenBank mRNA | AK175439.1 | - | 10 - 11 | 888 | 122 | |
AK176650.1 | - | 10 - 11 | 888 | 122 | ||
BT010880.1 | - | 10 - 11 | 877 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
