Assay Details
Gene Symbol: EFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084367.2 | NP_001077836.1 | 5 - 6 | 3059 | 72 | |
NM_106379.9 | NP_177854.6 | 5 - 6 | 3211 | 72 | ||
GenBank mRNA | EU014690.1 | - | 4 - 5 | 2924 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
