Assay Details
Gene Symbol: EFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084367.2 | NP_001077836.1 | 10 - 11 | 3534 | 104 | |
NM_106379.9 | NP_177854.6 | 10 - 11 | 3686 | 104 | ||
GenBank mRNA | EU014690.1 | - | 9 - 10 | 3399 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
