Assay Details
Gene Symbol: AT1G77550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106404.6 | NP_177879.3 | 13 - 14 | 2346 | 74 | |
GenBank mRNA | AK226662.1 | - | 13 - 14 | 2313 | 74 | |
BT014678.1 | - | 13 - 14 | 2261 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
