Assay Details
Gene Symbol: SHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198498.1 | NP_001185427.1 | 5 - 6 | 912 | 122 | |
NM_001334830.1 | NP_001322353.1 | 5 - 6 | 1010 | 122 | ||
NM_106501.5 | NP_177975.3 | 5 - 6 | 1010 | 122 | ||
GenBank mRNA | AK221532.1 | - | 5 - 6 | 900 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
