Assay Details
Gene Symbol: SRF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106547.3 | NP_178019.2 | 5 - 6 | 582 | 78 | |
GenBank mRNA | AY518290.1 | - | 5 - 6 | 582 | 78 | |
FJ708683.1 | - | 5 - 6 | 427 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
