Assay Details
Gene Symbol: MYB63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334873.1 | NP_001321204.1 | 1 - 2 | 380 | 94 | |
NM_106569.4 | NP_178039.1 | 2 - 3 | 519 | 94 | ||
GenBank mRNA | AK175344.1 | - | 2 - 3 | 281 | 94 | |
AY519572.1 | - | 2 - 3 | 267 | 94 | ||
BT033125.1 | - | 2 - 3 | 267 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
