Assay Details
Gene Symbol: AT3G10210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111856.3 | NP_566369.1 | 1 - 2 | 375 | 77 | |
GenBank mRNA | AY054242.1 | - | 1 - 2 | 238 | 77 | |
AY079116.1 | - | 1 - 2 | 229 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
