Assay Details
Gene Symbol: AT3G10210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111856.3 | NP_566369.1 | 2 - 3 | 643 | 63 | |
GenBank mRNA | AY054242.1 | - | 2 - 3 | 506 | 63 | |
AY079116.1 | - | 2 - 3 | 497 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
