Assay Details
Gene Symbol: GLY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111922.4 | NP_187696.1 | 1 - 2 | 279 | 63 | |
GenBank mRNA | AK228542.1 | - | 1 - 2 | 93 | 63 | |
AY052329.1 | - | 1 - 2 | 69 | 63 | ||
BT000849.1 | - | 1 - 2 | 40 | 63 | ||
Y08357.1 | - | 61 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
