Assay Details
Gene Symbol: GLY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111922.4 | NP_187696.1 | 2 - 3 | 411 | 83 | |
GenBank mRNA | AK228542.1 | - | 2 - 3 | 225 | 83 | |
AY052329.1 | - | 2 - 3 | 201 | 83 | ||
BT000849.1 | - | 2 - 3 | 172 | 83 | ||
Y08357.1 | - | 193 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
