Assay Details
Gene Symbol: GLY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111922.4 | NP_187696.1 | 3 - 4 | 568 | 114 | |
GenBank mRNA | AK228542.1 | - | 3 - 4 | 382 | 114 | |
AY052329.1 | - | 3 - 4 | 358 | 114 | ||
BT000849.1 | - | 3 - 4 | 329 | 114 | ||
Y08357.1 | - | 350 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
